Orders LinkContactUsLinkComponent
Concentrator
Koncentrator, opløsning, til brug med Tube-O-DIALYZER™, 125 mL
Katalog # 786-143
Leverandør:   G-Biosciences
Concentrator
Katalog # 786-143
Leverandør:  G-Biosciences
CAS nummer:  
Concentrator
Koncentrator, opløsning, til brug med Tube-O-DIALYZER™, 125 mL
CAS nummer:  
Price
1.100,00 kr
Log ind for tilgængelighed
1 * 125 mL
Product Details & Documents
The concentrator is a high molecular weight polymer which will not migrate across the dialysis membrane. The concentrator is placed against the dialysis membrane of the Tube-O-DIALYZER™, where it rapidly absorbs water from the sample and reduces the sample volume. The concentrator can be used with dialysis bags and other dialysis devices.
Specifikationer
Specifikationer
  • Pakn:
    125 mL
  • Beskrivelse:
    Koncentrator, opløsning, til brug med Tube-O-DIALYZER™
Specifikationer