Orders LinkContactUsLinkComponent
Blocking buffers, Protein-Free™
Blocking buffers, Protein-Free™
Protein-Free™ Blocking Buffer TBS with Tween® 20, 500 mL
Katalog # 786-663
Leverandør:   G-Biosciences
Blocking buffers, Protein-Free™
Katalog # 786-663
Leverandør:  G-Biosciences
CAS nummer:  
Blocking buffers, Protein-Free™
Protein-Free™ Blocking Buffer TBS with Tween® 20, 500 mL
CAS nummer:  
Price
1.170,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents
Protein-Free™ Blocking buffer is a formulation of non-protein agents that eliminate non-specific binding sites in ELISA, blotting, immunohistochemistry and other applications. The absence of protein from the Protein-Free™ Blocking buffer eliminates problems associated with traditional protein based blockers, such as cross-reactivity and interference from glycosylated proteins.
Protein-Free™ Blocking buffer is compatible with antibodies and avidin/biotin based systems and results in high signal to background ratios.
Protein-Free™ Blocking buffer is offered in four convenient, ready to use formats, with or without detergent. Protein-Free™ Blocking buffer is supplied either in TBS buffer (Tris-buffered saline at pH 7,5) or PBS buffer (phosphate-buffered saline at pH 7,5), as well as in separate formulations that contain Tween®-20 for improving blocking efficiencies.
Specifikationer
Specifikationer
  • Pakn:
    500 mL
  • Beskrivelse:
    Protein-Free™ Blocking Buffer TBS with Tween® 20