Orders LinkContactUsLinkComponent
Destain solutions
Destain solutions
Destain II (5% Methanol, 7% Acetic acid), 1 L
Katalog # 786-527
Leverandør:   G-Biosciences
Destain solutions
Katalog # 786-527
Leverandør:  G-Biosciences
CAS nummer:  
Destain solutions
Destain solutions
Destain II (5% Methanol, 7% Acetic acid), 1 L
CAS nummer:  
Price
kr610.00
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents
G-Biosciences' Destain I contains approximately 45% methanol and 9% glacial acetic acid. G-Biosciences' Destain II contains approximately 5% methanol and 7% acetic acid.
Specifikationer
Specifikationer
  • Pakn:
    1 L
  • Beskrivelse:
    Destain II (5% Methanol, 7% Acetic acid)