Orders LinkContactUsLinkComponent
Gel drying solution for protein gels
Gel drying solution for protein gels
Gel drying solution
Katalog # 786-685
Leverandør:   G-Biosciences
Gel drying solution for protein gels
Katalog # 786-685
Leverandør:  G-Biosciences
Gel drying solution for protein gels
Gel drying solution for protein gels
Gel drying solution
Price
1.150,00 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents
This gel drying solution effectively and evenly dries protein electrophoresis gels by the mechanism of passive evaporation and is suitable for permanent gel storage and densitometry.
  • Ready to use
  • Regulates rate of drying
  • Prevents cracking of the gel
  • Compatible with all drying membranes
Specifikationer
  • Beskrivelse:
    Gel drying solution
  • Seq:
    02