Solid phase extraction cartridges, HyperSep™ SCX
Patron, SCX, 40 - 60 µm, 75 ml, 20000 mg
Katalog # HYPE60108-857
Leverandør: Thermo Fisher Scientific
Solid phase extraction cartridges, HyperSep™ SCX
Katalog # HYPE60108-857
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
A strong cation exchange sorbent for extraction of charged basic compounds.
- Benzene ring provides potential for nonpolar interactions
- Low pKa value sorbent
Specifikationer
- Kolonne form:Patron
- Partikelstørrelse:40 - 60 µm
- Kapacitet:75 ml
- Seq:10
- Lejevægt:20000 mg
- Fase:SCX