Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
HyperSep™ Hypercarb™ SPE columns

HyperSep™ Hypercarb™ SPE columns

15 ml, 2000 mg
Katalog # 549-0627
Leverandør:   Thermo Fisher Scientific
HyperSep™ Hypercarb™ SPE columns
Katalog # 549-0627
HyperSep™ Hypercarb™ SPE columns
HyperSep™ Hypercarb™ SPE columns

15 ml, 2000 mg

Price
14.810,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents

Flat, 100% porous graphitic carbon (PGC) with selectivity for structurally similar compounds, offering separation of compounds with simple solvents. Hydrophobic reversed phase; normal phase adsorption; polar retention.

  • Provides total pH stability and the retention and separation of highly polar species where silica and resin columns fail
  • High batch-to-batch reproducibility
  • Strong retention properties allow use of low bed weights for concentrated extracts
  • No restriction on eluents
Specifikationer
  • Kapacitet:
    15 ml
  • Seq:
    07
  • Lejevægt:
    2000 mg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.