Eyewash stations, Duo
Product Details & Documents
Wall-mounted eyewash stations, made in hard plastic, for simple and quick first-aid in case of an eye injury.
- Easily wall-mountable
- Ensures quick and efficient eyewash
- Duo eye-cup for simultaneous washing of both eyes
- Supplied with pictograms and mirror
- Closed stations (boxes) are ideal for use in dirty and dusty areas
Eyewash station (with Eye Wash Duo):
Wall-mounted station containing 1×1000 ml Eye Wash Duo bottle, wall mounting device, pictogram and mirror. Especially for general eye washing at smaller work places.
Rinsing time: ~5 min
CombiStation Duo (with Eye Wash Duo + pH Neutral Duo):
Wall-mounted station containing 500 ml pH Neutral Duo and 1000 ml Eye Wash Duo, wall mounting device, pictogram and mirror. Especially suitable in places where there is a risk of eye injuries involving acids and alkalis.
Rinsing time: ~7 min (2 min with pH Neutral Duo and 5 min with Eye Wash Duo)
Combi Box Duo MAXI (with Eye Wash Duo + pH Neutral Duo):
Dust‐proof wall station containing 1×500 ml pH Neutral Duo and 1×1000 ml Eye Wash Duo, and a mirror inside the lid. Especially suitable for dusty production areas where there is a risk of eye injuries involving acids and alkali.
Rinsing time: ~7 min (2 min with pH Neutral Duo and 5 min with Eye Wash Duo)
Eyewash box Duo (with Eye Wash Duo):
Dust‐proof wall station containing 2×1000 ml Eye Wash Duo, and a mirror inside the lid. Especially suitable for dirty and dusty production areas and mobile work places.
Rinsing time: ~10 min.
Documents
Certifikater
Søg efter partinummerSpecifikationer
- Beskrivelse:CombiBox Duo med 1×1000 ml øjenskyllevæske i duoflaske og 1×500 ml pH-neutraliseringsvæske i duoflaske
- Seq:03