Orders LinkContactUsLinkComponent
Hematocrit tubes, PLASTICRIT™
Hematocrit tubes, PLASTICRIT™
Hematocrit Tubes, PLASTICRIT™ Plain, 75 mm
Katalog # DRUM8-000-7520/5
Leverandør:   DRUMMOND
Hematocrit tubes, PLASTICRIT™
Katalog # DRUM8-000-7520/5
Leverandør:  DRUMMOND
Hematocrit tubes, PLASTICRIT™
Hematocrit Tubes, PLASTICRIT™ Plain, 75 mm
Price
2.380,00 kr
Log ind for tilgængelighed
1 * 1.000 Items
Product Details & Documents
These plastic hematocrit tubes provide the identical capillary action as glass hematocrit tubes. They eliminate the possibility of contamination due to breakage with no compromise in accuracy.
  • Available as plain or heparinised
Complies to FDA, NIOSH, CDC, and OSHA safety recommendations.
Specifikationer
  • Beskrivelse:
    Hematocrit Tubes, PLASTICRIT™ Plain
  • Længde:
    75 mm
  • Seq:
    01