Cover glasses
High precision cover glasses, 24×50 mm, #1,5, Borosilikatglas
Katalog # 630-2187
Leverandør: MARIENFELD
Gå til:
Product Details & Documents
Cover slides suitable for fluorescence microscopy.
- Borosilicate glass D 263™ M of hydrolytic class 1
Recommended for objectives with high numerical aperture and resolution.
Documents
Certifikater
Søg efter partinummerSpecifikationer
- Antal:#1,5
- Beskrivelse:High precision cover glasses
- L×B:24×50 mm
- Materiale:Borosilikatglas
- Seq:03