Orders LinkContactUsLinkComponent
Control valves with GL thread, BOLA
Control valves with GL thread, BOLA
GL control valve with two-way straight bore Ø 6 mm, thread GL 18, PTFE
Katalog # BOHLE694-18
Leverandør:   Bohlender
Control valves with GL thread, BOLA
Katalog # BOHLE694-18
Leverandør:  Bohlender
Control valves with GL thread, BOLA
Control valves with GL thread, BOLA
GL control valve with two-way straight bore Ø 6 mm, thread GL 18, PTFE
Price
7.490,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Two-way valve with straight bore and two connections with GL thread completely made of PTFE.
  • Suitable for vacuum
  • Universal chemical resistance, the flowing product is only exposed to PTFE
Applications include for distributing liquids or gases, manual regulation for constant flow.
Specifikationer
  • Beskrivelse:
    GL control valve with two-way straight bore Ø 6 mm, thread GL 18, PTFE