Volumetric Flasks, Class A
10 ml, 14/23, 0,10 ± ml
Katalog # WITG3.664.00114.BL
Leverandør: witeg Labortechnik
Gå til:
Product Details & Documents
Borosilicate glass 3.3, with or without stopper.
- DE-M marked according to the German calibration law
- Inclusive batch certificate
Certifications: Tolerance according to DIN EN ISO 1042
Specifikationer
- NS:14/23
- Kapacitet:10 ml
- Seq:4
- Tolerance:0,10 ± ml