Inoculating loops and needles
Podeøser, 1 µl, hvid, 48/foliepakke
Catalog # SARS86.1567.050
Marketsource
Supplier: SARSTEDT
Gå til:
Product Details & Documents
PS, gamma sterilised. Time-saving, safe and easy to use. For laboratories with high workloads.
Specifikationer
- Pakket:48/foliepakke
- Kapacitet:1 µl
- Farve:hvid
- Beskrivelse:Podeøser