Replacement Erlenmeyer flask caps, Corning®
Ventileret låg, 43 mm, til 500 ml og 1 l kolbe (fremstilles kun på bestilling, minimumbestilling 5 kasser)
Katalog # 392-0292
Leverandør: Corning
Gå til:
Product Details & Documents
Made of PP.
Replacement caps for Corning® 500 ml, 2 and 3 litre Erlenmeyer flasks are available separately. They are supplied sterile and individually packaged.
Advarsel: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Specifikationer
- Beskrivelse:Ventileret låg, 43 mm, til 500 ml og 1 l kolbe (fremstilles kun på bestilling, minimumbestilling 5 kasser)
- Seq:01