Orders LinkContactUsLinkComponent
Inoculating loops and needles
1 µl, ufarvet
Katalog # KART2650
Leverandør:   KARTELL
Inoculating loops and needles
Katalog # KART2650
Leverandør:  KARTELL
Inoculating loops and needles
Inoculating loops and needles
1 µl, ufarvet
Price
1.280,00 kr
Log ind for tilgængelighed
1 * 2.000 Items
Product Details & Documents
High impact PS, sterile.
  • Choice of either 1 µl or 10 µl loop on one end and needle at the other
  • Sterilised by gamma irradiation
 : 20 per bag.
Specifikationer
  • Kapacitet:
    1 µl
  • Farve:
    ufarvet
  • Seq:
    01