Orders LinkContactUsLinkComponent
Shelving systems, direct wall mounts
Shelving systems, direct wall mounts
Shelf, double, 1,4 kg
Katalog # METO2WD18S
Leverandør:   Metro International
Shelving systems, direct wall mounts
Katalog # METO2WD18S
Leverandør:  Metro International
Shelving systems, direct wall mounts
Shelving systems, direct wall mounts
Shelf, double, 1,4 kg
Price
2.100,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Direct wall mount brackets allow the installation of wire or solid shelves directly onto a wall surface. Each brackets consists of one shelf support and mounting plate, with two shelf collar caps. Use single support at shelf ends and double support for adjoining shelves.
Specifikationer
  • Beskrivelse:
    Shelf, double
  • Seq:
    04
  • Vægt:
    1,4 kg