Shelving systems, direct wall mounts
Shelf, double, 1,4 kg
Katalog # METO2WD18S
Leverandør: Metro International
Gå til:
Product Details & Documents
Direct wall mount brackets allow the installation of wire or solid shelves directly onto a wall surface. Each brackets consists of one shelf support and mounting plate, with two shelf collar caps. Use single support at shelf ends and double support for adjoining shelves.
Specifikationer
- Beskrivelse:Shelf, double
- Seq:04
- Vægt:1,4 kg