Læselup, easyPOCKET
easyPOCKET, 4×, 50×45 mm
630-1321
These pocket magnifiers provide magnification and illumination in a credit card format.
- Ultra-flat and lightweight design
- Diffractive aspheric PXM® lightweight lens
- SMD LED illumination, which is switched on when you slide the lens out of its case
- Easy handling - No bulb change required
: Supplied with two button cells.
- Forstørrelse:4×
- Beskrivelse:easyPOCKET
- Seq:02
- B×D:50×45 mm