Orders LinkContactUsLinkComponent
Einhorn's saccharometer
Einhorn's saccharometer
Ekstra fortyndingsslange, brun inddeling, mærke ved 10 ml
Catalog # HECH41370020
Einhorn's saccharometer
Catalog # HECH41370020
Einhorn's saccharometer
Einhorn's saccharometer
Ekstra fortyndingsslange, brun inddeling, mærke ved 10 ml
Price
38,20 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Fermentation saccharometers are used for the estimation of sugar in urine and diabetes diagnostic.
The carbon dioxide created in the fermenting process would rise to the top of the closed tube and force the level of liquid down.
The percentage of sugar could be read off the graduated scale on the closed tube side of the apparatus.
Specifications
  • Beskrivelse:
    Ekstra fortyndingsslange, brun inddeling, mærke ved 10 ml