o-Cresol 99%
Glass bottle, 100 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: o-Methylphenol
- CAS nummer:95-48-7
- Formel:C₇H₈O
- MDL nummer:MFCD00002226
- Molecular Weight:108.14 g/mol
- Merck Index:13,02604
- Kogende Pt:191 °C (760 mmHg)
- Smeltende Pt:30…32 °C
- Tæthed:1,04 g/cm³
- Flash Pt:81 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:202-423-8
- FN:3455
- ADR:6.1,II
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Colorless to amber Low melting solid or liquid (May darken on storage)
- Infrared spectrum:Conforms
- Water:≤0.2 % (K.F.)
- GC:≥98.5 %
Specifikationer
- Pakning:Glass bottle
- Pakn:100 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



