Storage bags
Storage bags, transparent, with ground weld., 0,04 mm, 240×380 mm
Katalog # SEMA2448
Leverandør: SEMADENI
Gå til:
Product Details & Documents
Storage bags in transparent LDPE, with bottom weld.
They are also suitable for liquids but without side weld.
Emballering: Minimum quantity:100 pieces
Specifikationer
- Beskrivelse:Storage bags, transparent, with ground weld.
- Dimensioner:240×380 mm
- Seq:06
- Tykkelse:0,04 mm