Orders LinkContactUsLinkComponent
Glasmikrofiber-forfiltre
Glasmikrofiber-forfiltre
79 mm
Katalog # 734-2300
Leverandør:   Corning
Glasmikrofiber-forfiltre
Katalog # 734-2300
Leverandør:  Corning
Glasmikrofiber-forfiltre
79 mm
Price
836,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
Glass fiber filters are used a depth filter for prefiltration of solutions.
They have very high particle loading capacity and are ideal for prefiltering dirty solutions and difficult to filter biological fluids such as sera.
Documents
Specifikationer
  • Størrelse:
    79 mm