Orders LinkContactUsLinkComponent
Glasmikrofiber-forfiltre
63 mm
Katalog # 734-2299
Leverandør:   Corning
Glasmikrofiber-forfiltre
Katalog # 734-2299
Leverandør:  Corning
Glasmikrofiber-forfiltre
63 mm
Price
716,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
Glass fiber filters are used a depth filter for prefiltration of solutions.
They have very high particle loading capacity and are ideal for prefiltering dirty solutions and difficult to filter biological fluids such as sera.
Specifikationer
  • Størrelse:
    63 mm