Glasmikrofiber-forfiltre
43 mm
Leverandør: Corning
Gå til:
Product Details & Documents
Glass fiber filters are used a depth filter for prefiltration of solutions.
They have very high particle loading capacity and are ideal for prefiltering dirty solutions and difficult to filter biological fluids such as sera.
Documents
Certifikater
Søg efter partinummerSpecifikationer
- Størrelse:43 mm