Orders LinkContactUsLinkComponent
Immersion coolers
Immersion coolers
140 mm, 29/32 NS
Katalog # WITG1.786.229.S
Leverandør:   witeg Labortechnik
Immersion coolers
Katalog # WITG1.786.229.S
Leverandør:  witeg Labortechnik
Immersion coolers
Immersion coolers
140 mm, 29/32 NS
Price
823,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Borosilicate glass 3,3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For immersion into distillation assemblies etc. Can be used to regulate refluxing by adjusting the flow of air or water through it. Water connection with two glass olives.
Specifikationer
  • Konusstr.:
    29/32 NS
  • Kondenserlængde:
    140 mm