Orders LinkContactUsLinkComponent
Test tubes, flat bottom, without rim, heavy wall
Test tubes, flat bottom, without rim, heavy wall
65 mm, 15 mm
Katalog # HECH42779065
Test tubes, flat bottom, without rim, heavy wall
Katalog # HECH42779065
Test tubes, flat bottom, without rim, heavy wall
Test tubes, flat bottom, without rim, heavy wall
65 mm, 15 mm
Price
242,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
Made of AR-Glas® (soda glass).
  • Heavy wall
  • Flat bottom
  • Good chemical and physical properties
Documents
Specifikationer
  • Udv.Ø:
    15 mm
  • Længde:
    65 mm
  • Seq:
    02