Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
HEPES ≥99.5% suitable for buffer solutions

HEPES ≥99.5% suitable for buffer solutions

Plastic drum, open-head, 25 kg
Katalog # A1069.9025BW
Leverandør:   PanReac AppliChem
HEPES ≥99.5% suitable for buffer solutions
Katalog # A1069.9025BW
Leverandør:  PanReac AppliChem
CAS nummer:  7365-45-9
HEPES ≥99.5% suitable for buffer solutions
HEPES ≥99.5% suitable for buffer solutions

Plastic drum, open-head, 25 kg

CAS nummer:  7365-45-9
Price
Spørg for pris
Log ind for tilgængelighed
1 * 25 kg
Product Details & Documents
Synonymer:  HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid
  • CAS nummer:7365-45-9
  • Formel:C₈H₁₈N₂O₄S
  • MDL nummer:MFCD00006158
  • Molecular Weight:238.31 g/mol
  • Merck Index:13,04672
  • Smeltende Pt:210…215 °C
  • Tæthed:1,448 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:230-907-9
Specifikationer

Specifikationstestresultater

  • Assay (titr.):
    min. 99.5 %
  • pH (1 %; H2O):
    4.7 - 6.0
  • Heavy metals (as Pb):
    max. 0.001 %
  • Water:
    max. 0.5 %
  • Chloride:
    max. 0.05 %
  • Sulfate:
    max. 0.05 %

Specifikationer

  • Pakning:
    Plastic drum, open-head
  • Pakn:
    25 kg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.