Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillaries for LC systems
Capillaries, pre-swaged, male to male, 320 mm, I.Ø 0,17 mm
  HEWL5067-1571
Capillaries for LC systems
  HEWL5067-1571
Capillaries for LC systems
Capillaries for LC systems
Capillaries, pre-swaged, male to male, 320 mm, I.Ø 0,17 mm
717,00 kr
1 * 1 Items
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
  • Beskrivelse:
    Capillaries, pre-swaged, male to male, 320 mm, I.Ø 0,17 mm
  • Seq:
    54