Bucket, Rectangular
Bucket with colour coding clips, grå, 6 L
Katalog # 129-0458
Leverandør: FHP VILEDA SCS
Gå til:
Product Details & Documents
Polypropylene bucket with stainless steel handle. For use on the CE Bucket Trolley or the CE pre-prepared trolley.
- Liquid measurement lines molded inside of bucket
- Color coding clips included to allow differentiation in colours of buckets (red, blue, green, yellow)
- Autoclavable
Highest performance for the best contamination control.
Specifikationer
- Kapacitet:6 L
- Farve:grå
- Beskrivelse:Bucket with colour coding clips
- Seq:01