Orders LinkContactUsLinkComponent
Eco caps
Eco cap kit, with 2 l safety coated glass bottle
Katalog # JOURJR-9000-0014
Leverandør:   VICI JOUR RESEARCH
Eco caps
Katalog # JOURJR-9000-0014
Leverandør:  VICI JOUR RESEARCH
Eco caps
Eco cap kit, with 2 l safety coated glass bottle
Price
2.130,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The Eco Cap consists of a three-hole ETFE insert with a polyethylene collar.
  • Compatible with all standard GL 45 thread bottles
  • Also available as bottle kit
There is no threading for fittings on the inside of the tubing ports, the tubing can be easily inserted.
 : The Eco Cap is supplied with one cap, polypropylene male Luer plug. The kit also includes safety-coated glass bottle with a option of 1 or 2 L.
Specifikationer
  • Beskrivelse:
    Eco cap kit, with 2 l safety coated glass bottle
  • Seq:
    03