Orders LinkContactUsLinkComponent
Double powder spatulas, micro
Double powder spatulas, micro
150 mm
Catalog # HAMMHSN011-15
Supplier:   HAMMACHER KARL
Double powder spatulas, micro
Catalog # HAMMHSN011-15
Supplier:  HAMMACHER KARL
Double powder spatulas, micro
Double powder spatulas, micro
150 mm
Price
87,40 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Flexible, double-ended micro powder spatulas made of stainless steel.
Specifications
  • Længde:
    150 mm
  • Seq:
    01