Orders LinkContactUsLinkComponent
Milk butyrometer
Milk butyrometer
Butyrometer for milk, 0 - 5% : 0,1
Katalog # GERB3151-C
Leverandør:   GERBER FUNKE
Milk butyrometer
Katalog # GERB3151-C
Leverandør:  GERBER FUNKE
Milk butyrometer
Milk butyrometer
Butyrometer for milk, 0 - 5% : 0,1
Price
249,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Borosilicate glass.
  • With or without stopper
Available with a declaration of conformity per batch and type (in German only).
Specifikationer
  • Beskrivelse:
    Butyrometer for milk, 0 - 5% : 0,1
  • Seq:
    14
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Stopper, rubber, conical, for all measuring method butyrometers
    Stopper, rubber, conical, for all measuring method butyrometers
    614-2028
    Log ind for tilgængelighed
    1 * 100 Items - 360,00 kr