Cover slides
24×60 mm, 0,15 mm
Leverandør: DELTALAB
Gå til:
Product Details & Documents
Clear and pre-washed glass cover slides dedicated to sample and lens protection.
Delivery information: Supplied in cases of one or two hundred.
Documents
Certifikater
Søg efter partinummerSpecifikationer
- L×B:24×60 mm
- Seq:03
- Tykkelse:0,15 mm