Tubes for ParasiTrap®
Filled tubes I and II, integrated filters, flotation fluid and accessories
8488FLOT/50
: Biosepar
- Beskrivelse:Filled tubes I and II, integrated filters, flotation fluid and accessories
- Seq:08