Interfaces
Com.G@te®, internal for Petite Fleur®, Grande Fleur®, Unichiller® with Pilot ONE, Ministats®, CC®-300BX to CC®-906w
Katalog # HUBE31217
Leverandør: HUBER
Gå til:
Product Details & Documents
Com.G@te®. The interfaces are integrated RS232 (bi-directional), RS485 (bi-directional), ECS external control signal, volt free contact, AIF analogue interface 0/4 to 20 mA or 0 to 10 V (bi-directional). Connections comply with NAMUR standard.
Specifikationer
- Beskrivelse:Com.G@te®, internal for Petite Fleur®, Grande Fleur®, Unichiller® with Pilot ONE, Ministats®, CC®-300BX to CC®-906w
- Seq:01