Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

ISE calibration standards

Nitrate ISE standard solution, 1000 mg/l, 1000 mg/l as NO₃-, 500 mL
Katalog # METT51344779
Leverandør:   Mettler - Toledo
ISE calibration standards
Katalog # METT51344779
Leverandør:  Mettler - Toledo
CAS nummer:  
ISE calibration standards
ISE calibration standards

Nitrate ISE standard solution, 1000 mg/l, 1000 mg/l as NO₃-, 500 mL

CAS nummer:  
Price
1.160,00 kr
Log ind for tilgængelighed
1 * 500 mL
Specifikationer

Specifikationer

  • Koncentration:
    1000 mg/l as NO₃-
  • Pakn:
    500 mL
  • Beskrivelse:
    Nitrate ISE standard solution, 1000 mg/l

Specifikationer