Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
ISE calibration standards

ISE calibration standards

Silver ISE standard solution, 1000 mg/l, 1000 mg/l as Ag, 500 mL
Katalog # METT51344770
Leverandør:   Mettler - Toledo
ISE calibration standards
Katalog # METT51344770
Leverandør:  Mettler - Toledo
CAS nummer:  
ISE calibration standards
ISE calibration standards

Silver ISE standard solution, 1000 mg/l, 1000 mg/l as Ag, 500 mL

CAS nummer:  
Price
1.480,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents
Specifikationer

Specifikationer

  • Koncentration:
    1000 mg/l as Ag
  • Pakn:
    500 mL
  • Beskrivelse:
    Silver ISE standard solution, 1000 mg/l

Specifikationer