Orders LinkContactUsLinkComponent
Large hydrometers
Large hydrometers
10 - 20 % mas, 0,1 % mas, 650 mm
Catalog # AMARH847302
Supplier:   Amarell
Large hydrometers
Catalog # AMARH847302
Supplier:  Amarell
Large hydrometers
10 - 20 % mas, 0,1 % mas, 650 mm
Price
1.490,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Large hydrometers specially suited for breweries.
Specifications
  • Længde:
    650 mm
  • Seq:
    02
  • Inddeling:
    0,1 % mas
  • Område:
    10 - 20 % mas