Screw Caps
GL 18
Catalog # HECH41326018
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
PP, sort.
Very good chemical resistance
- Heat-resistant up to 110 °C
- Only suitable for bottles 215-2814 to 215-2819
Specifications
- Gevind:GL 18
- Seq:1