Orders LinkContactUsLinkComponent
Cell viability assay kits
Cell viability assay kits
Resazurin Cell Viability Assay kit, 10 000 assays (100 ml), 1 KIT
Katalog # 30025-2
Leverandør:   Biotium
Cell viability assay kits
Katalog # 30025-2
Leverandør:  Biotium
CAS nummer:  
Cell viability assay kits
Resazurin Cell Viability Assay kit, 10 000 assays (100 ml), 1 KIT
CAS nummer:  
Price
3.890,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents
MTT, XTT, and resazurin are reduced by mitochondrial metabolic activity to yield coloured or fluorescent products, and thus are useful for and assaying cell viability and quantitating cell number. MTT and XTT are reduced to coloured formazin salts that can be measured by absorbance. MTT generates an insoluble formazin salt, requiring cell lysis before the absorbance can be measured, while XTT does not require cell lysis for measurement. Resazurin is a non-fluorescent blue dye that is reduced to the pink fluorescent compound resorufin, which can be measured by fluorescence or absorbance.
Documents
Specifications
Specifications
  • Pakn:
    1 KIT
  • Beskrivelse:
    Resazurin Cell Viability Assay kit, 10 000 assays (100 ml)