Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
ISE calibration standards

ISE calibration standards

Chloride ISE standard solution, 1000 mg/l, 1000 mg/l as Cl, 500 mL
Catalog # METT51344772
Supplier:   Mettler - Toledo
ISE calibration standards
Catalog # METT51344772
Supplier:  Mettler - Toledo
CAS Number:  
ISE calibration standards
ISE calibration standards

Chloride ISE standard solution, 1000 mg/l, 1000 mg/l as Cl, 500 mL

CAS Number:  
Price
1.160,00 kr
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Koncentration:
    1000 mg/l as Cl
  • Pakn:
    500 mL
  • Beskrivelse:
    Chloride ISE standard solution, 1000 mg/l

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.