Orders LinkContactUsLinkComponent
Cell viability assay kits
Resazurin Cell Viability Assay kit, 2500 assays (25 ml), 1 KIT
Katalog # 30025-1
Leverandør:   Biotium
Cell viability assay kits
Katalog # 30025-1
Leverandør:  Biotium
CAS nummer:  
Cell viability assay kits
Resazurin Cell Viability Assay kit, 2500 assays (25 ml), 1 KIT
CAS nummer:  
Price
1.640,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
MTT, XTT, and resazurin are reduced by mitochondrial metabolic activity to yield coloured or fluorescent products, and thus are useful for and assaying cell viability and quantitating cell number. MTT and XTT are reduced to coloured formazin salts that can be measured by absorbance. MTT generates an insoluble formazin salt, requiring cell lysis before the absorbance can be measured, while XTT does not require cell lysis for measurement. Resazurin is a non-fluorescent blue dye that is reduced to the pink fluorescent compound resorufin, which can be measured by fluorescence or absorbance.
Specifikationer
Specifikationer
  • Pakn:
    1 KIT
  • Beskrivelse:
    Resazurin Cell Viability Assay kit, 2500 assays (25 ml)
Specifikationer