Orders LinkContactUsLinkComponent
VWR® MaximumSF, Kitler til renrum
VWR® MaximumSF, Kitler til renrum
hvid, XXL, 100 cm
Katalog # 113-1142
Leverandør:   VWR Collection
VWR® MaximumSF, Kitler til renrum
Katalog # 113-1142
Leverandør:  VWR Collection
VWR® MaximumSF, Kitler til renrum
VWR® MaximumSF, Kitler til renrum
hvid, XXL, 100 cm
Price
1.050,00 kr
Log ind for tilgængelighed
1 * 25 Items
Product Details & Documents
VWR® Maximum SF lab coats are the cleanest and most durable within the entire VWR® clothing line. Made from spunbonded polypropylene fabric (SPP) and coated with breathable polyethylene film, these garments have been designed to optimise barrier protection, cleanliness, and comfort properties.
  • Tapered collar, inset sleeves, snap closure, elasticated cuffs and three pockets
  • Superior fluid and particulate barrier
  • Lowest level of particle shedding: Level I Helmke drum classification
  • Light chemical splash resistance
  • Improved breathability to ensure user comfort
  • Soft, cloth-like fabric with maximum tear strength
Testet og fremstillet i ISO-certificerede omgivelser under streng proceskontrol. Lot-kontrolleret og valideret af et uafhængigt testlaboratorium.
Specifikationer
  • Tøj størrelse:
    XXL
  • Farve:
    hvid
  • Længde:
    100 cm
  • Seq:
    09
  • Cat. No.:
    113-1142
  • Outfit applikationer:
    Cleanroom|Laboratory
  • Lukkestørrelse:
    Med trykknapper
  • Lommer:
    med lommer