Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
VWR® Advanced, Kitler, SPP

VWR® Advanced, Kitler, SPP

hvid, 3XL, 105 cm
Katalog # 113-1150
Leverandør:   VWR Collection
VWR® Advanced, Kitler, SPP
Katalog # 113-1150
Leverandør:  VWR Collection
VWR® Advanced, Kitler, SPP
VWR® Advanced, Kitler, SPP

hvid, 3XL, 105 cm

Price
1.030,00 kr
Log ind for tilgængelighed
1 * 25 Items
Product Details & Documents

VWR® Advanced SF lab coats are manufactured from spunbonded polypropylene fabric coated with breathable polyethylene film.

  • Tapered collar, inset sleeves, snap closure, elasticated cuffs and three pockets
  • Durable fabric with good breathability and water transmission rate
  • Significant fluid and particle barrier
  • Particle shedding: Level II Helmke drum classification
  • Soft, cloth-like fabric

Testet og fremstillet i ISO-certificerede omgivelser under streng proceskontrol. Lot-kontrolleret og valideret af et uafhængigt testlaboratorium.

Specifikationer
  • Tøj størrelse:
    3XL
  • Farve:
    hvid
  • Længde:
    105 cm
  • Seq:
    06
  • Køn:
    Unisex
  • Cat. No.:
    113-1150
  • Outfit applikationer:
    Cleanroom|Laboratory
  • Lukkestørrelse:
    Med trykknapper
  • Lommer:
    med lommer
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.