Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Multiple distributors for bottles , BOLA

Multiple distributors for bottles , BOLA

S 40, 2 × max. 8 mm, 2 × GL 14
Katalog # BOHLD624-04
Leverandør:   Bohlender
Multiple distributors for bottles , BOLA
Katalog # BOHLD624-04
Leverandør:  Bohlender
Multiple distributors for bottles , BOLA
Multiple distributors for bottles , BOLA

S 40, 2 × max. 8 mm, 2 × GL 14

Price
1.940,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

PTFE.

  • Very good chemical resistance
  • Temperature resistant from -200 to +250 °C
  • Suitable for vacuum applications

For the removal or filling of aggressive or pure liquids. For the connection of hoses, tubes and probes into vessels. Hoses with an external diameter of max. 8 mm can be inserted through the stopcocks.

Specifikationer
  • Gevind:
    S 40
  • Rør udv. Ø:
    2 × max. 8 mm
  • Hals:
    2 × GL 14
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.