Orders LinkContactUsLinkComponent
Propper, diamantslebne
Propper, diamantslebne
24/29
Catalog # 217-0804
Supplier:   DWK Life Sciences
Propper, diamantslebne
Catalog # 217-0804
Supplier:  DWK Life Sciences
Propper, diamantslebne
Propper, diamantslebne
24/29
Price
2.540,00 kr
Log in for availability
1 * 100 Items
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Hexagonal
  • Flat
  • Diamond-ground
Specifications
  • NS:
    24/29
  • Seq:
    10