Robot-spidser til Agilent
70 µl, +, -, 45,62 mm, Transparent, Agilent™ VPrep® and Bravo®, 384-head, 10 racks of 384 tips
Katalog # 736-0413
Leverandør: BIOTIX
Gå til:
Product Details & Documents
Biotix robotic tips for Agilent™ VPrep® and Bravo® platforms are engineered to ensure complete compatibility, requiring no changes in programming scripts in the platform's liquid handling software suite.
- Made of medical-grade virgin PP
- Tips are BioReady™ certified to be free of RNase, DNase, and Endotoxins (pyrogens)
- Available as standard or filtered tips
Manufactured to the highest engineering standards for robotic systems, the ultra-straight tips target the centre of microplate wells precisely, delivering exceptional accuracy and precision without risk of jamming.
Specifikationer
- Pakket:10 racks of 384 tips
- Steril:+
- Volumen:70 µl
- Længde:45,62 mm
- Seq:03
- Spits farge:Transparent
- Til brug med:Agilent™ VPrep® and Bravo®, 384-head
- Filter:-