Nessler cylinders
Nessler cylinder 100 ml, low form, with optically plane bottom, circular mark at 100 ml, Ø×L: 34×180 mm
Katalog # WITG2.151.100
Leverandør: witeg Labortechnik
Gå til:
Product Details & Documents
Colour comparison cylinders according to Nessler for comparators, with optically plane bottom.
Specifikationer
- Beskrivelse:Nessler cylinder 100 ml, low form, with optically plane bottom, circular mark at 100 ml, Ø×L: 34×180 mm
- Seq:05