Orders LinkContactUsLinkComponent
Phosphate standard solutions
Phosphate standard solution, pack of 20×2 ml ampoules (NIST), 50 mg/l as PO4, 20
Katalog # HACH17120H
Leverandør:   Hach
Phosphate standard solutions
Katalog # HACH17120H
Leverandør:  Hach
CAS nummer:  
Phosphate standard solutions
Phosphate standard solutions
Phosphate standard solution, pack of 20×2 ml ampoules (NIST), 50 mg/l as PO4, 20
CAS nummer:  
Price
608,00 kr
Log ind for tilgængelighed
1 * 20 Items
Product Details & Documents
Standard solution for accuracy checks in phosphate (phosphorus) determinations.
Specifikationer
Specifikationer
  • Koncentration:
    50 mg/l as PO<sub>4</sub>
  • Pakn:
    20
  • Beskrivelse:
    Phosphate standard solution, pack of 20×2 ml ampoules (NIST)
Specifikationer