Orders LinkContactUsLinkComponent
VWR®, Cell Holders
VWR®, Cell Holders
Cell holder, solid sample holder (δ 1,5 - 4 mm; 1-position; for single beam and split beam beam only; sample size (L*W): 120*80 mm)
Catalog # 634-6011
Supplier:   VWR Collection
VWR®, Cell Holders
Catalog # 634-6011
Supplier:  VWR Collection
VWR®, Cell Holders
VWR®, Cell Holders
Cell holder, solid sample holder (δ 1,5 - 4 mm; 1-position; for single beam and split beam beam only; sample size (L*W): 120*80 mm)
Price
2.600,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Beskrivelse:
    Cell holder, solid sample holder (δ 1,5 - 4 mm; 1-position; for single beam and split beam beam only; sample size (L*W): 120*80 mm)
  • Seq:
    11