Orders LinkContactUsLinkComponent
Chemo protective aprons, BioClean-C™
Chemo protective aprons, BioClean-C™
blå, L
Katalog # 113-8149
Leverandør:   Nitritex
Chemo protective aprons, BioClean-C™
Katalog # 113-8149
Leverandør:  Nitritex
Chemo protective aprons, BioClean-C™
Chemo protective aprons, BioClean-C™
blå, L
Price
3.030,00 kr
Log ind for tilgængelighed
1 * 50 Items
Product Details & Documents
These disposable aprons are made of CleanTough™ fabric, a spunbonded non woven PP fabric laminated with a PE film.
  • Processed to ensure ISO Class 4 compatibility
  • Lightweight, low linting and hard-wearing CleanTough™ fabric
  • Adjustable neck fastening
  • Tie tapes at the waist
The aprons provide protection against fine sprays and particles as well as against a range of chemotherapy drugs.
Colour: Blue
 : EN ISO 13034:2005 +A1:2010 (Type PB [6]), EN 14325, EN 530, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2; Cat. III
Manufactured in a facility holding ISO 9001:2015 and (EU) 2016/425 Module D certifications.
Sterile models: Sterilised in accordance with ISO 11137-1:2006.
 : One piece per sealed inner bag; one sealed inner bag per sealed outer bag; 50 outer bags per carton liner; one carton liner per carton (50 pieces).
Documents
Specifikationer
  • Tøj størrelse:
    L
  • Farve:
    blå
  • Seq:
    03
  • Cat. No.:
    113-8149
  • Outfit applikationer:
    Medical|Clinical