Counting Chambers, Neubauer
Counting chamber with double ruling, Neubauer, 0,02 mm
Katalog # HECH92044102702
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
These counting chambers are designed for blood tests and urine analysis.
- Counting erythrocytes corresponds exactly to Thoma method
- Counting leucocytes in five large squares
- Evaluation just as indicated for Thoma ruling
Consisting of nine large squares – The central square shows the Thoma ruling and the other eight squares are subdivided.
Documents
HECH40441702_TDS_EN
Datablad
Specifikationer
- Kammerdybde:0,02 mm
- Beskrivelse:Counting chamber with double ruling, Neubauer
- Seq:03